Assay Details
Gene Symbol: Sh2d1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033499.1 | NP_001028671.1 | 2 - 3 | 202 | 128 | |
GenBank mRNA | DQ131181.1 | - | 2 - 3 | 202 | 128 | |
DQ140178.1 | - | 2 - 3 | 202 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine phosphorylation
regulation of natural killer cell activation
negative regulation of natural killer cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of peptidyl-tyrosine phosphorylation
protein binding
peptidyl-tyrosine phosphorylation
regulation of natural killer cell activation
negative regulation of natural killer cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of peptidyl-tyrosine phosphorylation
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20821404 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20821405 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20821409 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072616 | NM_001033499.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072616 | DQ131181.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072616 | DQ140178.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | AK148080.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | AK165742.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | AK171901.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | BC018389.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | BC022172.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | D67067.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555418 | NM_019649.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144889 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299605 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561514 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640544 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320918 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129794 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756230 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821331 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28752 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5845 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24575 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149028 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072616 | NM_001033499.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072616 | DQ131181.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072616 | DQ140178.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04351415 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04351390 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00248909 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04351415 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04351390 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20821409 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20821405 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20821404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00779519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00847227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00691091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00556415 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00305118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01122263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00779526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00261568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00318224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821404 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01135743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20821294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00234632 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10351476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351475 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351474 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10351473 | - | - |
Affymetrix Mouse Clariom S Array | PSR0100012829.mm.1 | NM_001033499.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012829.mm.1 | NM_001033499.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012829.mm.1 | TC0100001567.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100012830.mm.1 | NM_001033499.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012830.mm.1 | NM_001033499.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012830.mm.1 | TC0100001567.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100012831.mm.1 | NM_001033499.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012831.mm.1 | NM_001033499.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012831.mm.1 | TC0100001567.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100012832.mm.1 | NM_001033499.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012832.mm.1 | NM_001033499.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012832.mm.1 | TC0100001567.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 767824 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257907 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2915800 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2915800 | - | - |
