Assay Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030732.3 | NP_109657.2 | 14 - 15 | 1829 | 73 | |
| GenBank mRNA | AF268195.1 | - | 14 - 15 | 1610 | 73 | |
| AK029595.1 | - | 14 - 15 | 1628 | 73 | ||
| AK033347.1 | - | 14 - 15 | 1699 | 73 | ||
| AK036064.1 | - | 15 - 16 | 1940 | 73 | ||
| AK136255.1 | - | 14 - 15 | 1651 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbl1xr1
Genomic Map