Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
NM_001243062.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
NM_001243062.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
NM_001243063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
NM_001243063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
NM_009803.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
NM_009803.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012934.mm.1 |
TC0100001579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
NM_001243062.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
NM_001243062.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
NM_001243063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
NM_001243063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
NM_009803.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
NM_009803.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012935.mm.1 |
TC0100001579.mm.1 |
MTA TC |