| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100012637.mm.1 |
NM_001164528.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012637.mm.1 |
NM_001164528.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012637.mm.1 |
TC0100001533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100012636.mm.1 |
NM_001164528.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012636.mm.1 |
NM_001164528.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012636.mm.1 |
TC0100001533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100012638.mm.1 |
NM_001164528.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012638.mm.1 |
NM_001164528.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012638.mm.1 |
TC0100001533.mm.1 |
MTA TC |