Assay Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019673.2 | NP_062647.2 | 3 - 4 | 588 | 95 | |
| GenBank mRNA | AF041476.1 | - | 3 - 4 | 408 | 95 | |
| AK002724.1 | - | 3 - 4 | 384 | 95 | ||
| AK090154.1 | - | 3 - 4 | 407 | 95 | ||
| AK167905.1 | - | 3 - 4 | 402 | 95 | ||
| AK169278.1 | - | 3 - 4 | 407 | 95 | ||
| BC001994.1 | - | 3 - 4 | 376 | 3491205 | 95 | |
| BC094498.1 | - | 3 - 4 | 404 | 4209491 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Actl6a
Genomic Map