Assay Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019673.2 | NP_062647.2 | 4 - 5 | 684 | 82 | |
| GenBank mRNA | AF041476.1 | - | 4 - 5 | 504 | 82 | |
| AK002724.1 | - | 4 - 5 | 480 | 82 | ||
| AK090154.1 | - | 4 - 5 | 503 | 82 | ||
| AK167905.1 | - | 4 - 5 | 498 | 82 | ||
| AK169278.1 | - | 4 - 5 | 503 | 82 | ||
| BC001994.1 | - | 4 - 5 | 472 | 3491205 | 82 | |
| BC094498.1 | - | 4 - 5 | 500 | 4209491 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Actl6a
Genomic Map