Assay Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019673.2 | NP_062647.2 | 6 - 7 | 884 | 81 | |
| GenBank mRNA | AK002724.1 | - | 6 - 7 | 680 | 81 | |
| AK090154.1 | - | 6 - 7 | 703 | 81 | ||
| AK167905.1 | - | 6 - 7 | 698 | 81 | ||
| AK169278.1 | - | 6 - 7 | 703 | 81 | ||
| BC001994.1 | - | 6 - 7 | 672 | 3491205 | 81 | |
| BC094498.1 | - | 6 - 7 | 700 | 4209491 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Actl6a
Genomic Map