Assay Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK167905.1 | - | 9 - 10 | 960 | 103 | |
| BC001994.1 | - | 9 - 10 | 934 | 3491205 | 103 | |
| BC094498.1 | - | 9 - 10 | 962 | 4209491 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actl6a
Genomic Map