Assay Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163516.2 | NP_001156988.1 | 6 - 7 | 961 | 124 | |
| NM_001289505.1 | NP_001276434.1 | 8 - 9 | 942 | 124 | ||
| NM_021483.4 | NP_067458.2 | 8 - 9 | 1071 | 124 | ||
| GenBank mRNA | AB032591.1 | - | 8 - 9 | 1066 | 124 | |
| AK039764.1 | - | 8 - 9 | 941 | 124 | ||
| AK044552.1 | - | 8 - 9 | 1199 | 124 | ||
| AK083141.1 | - | 6 - 7 | 853 | 124 | ||
| HQ646186.1 | - | 9 - 10 | 1290 | 124 | ||
| HQ646187.1 | - | 8 - 9 | 1224 | 124 | ||
| HQ646188.1 | - | 7 - 8 | 1119 | 124 | ||
| HQ646189.1 | - | 7 - 8 | 1152 | 124 | ||
| HQ646190.1 | - | 6 - 7 | 1047 | 124 | ||
| HQ646191.1 | - | 8 - 9 | 1880 | 124 | ||
| HQ646192.1 | - | 9 - 10 | 1294 | 124 | ||
| HQ646193.1 | - | 9 - 10 | 1200 | 124 | ||
| HQ646194.1 | - | 8 - 9 | 1095 | 124 | ||
| HQ646195.1 | - | 9 - 10 | 1068 | 124 | ||
| HQ646196.1 | - | 8 - 9 | 963 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083141.1 transcripts.
Set Membership: |
|

Pex5l
Genomic Map