Assay Details
| Gene Symbol: Usp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013024.2 | NP_001013042.1 | 12 - 13 | 1615 | 60 | |
| GenBank mRNA | AK135141.1 | - | 12 - 13 | 1667 | 60 | |
| BC090999.1 | - | 12 - 13 | 1615 | 6754549 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp13
Genomic Map