Assay Details
| Gene Symbol: Gm20557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030119.1 | - | 14 - 15 | 2174 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | enhancer of rudimentary homolog pseudogene | 1700018B24Rik | -745.4kb | Anti-sense | Chr. 3 - 48605731 - 48609100 on NCBI Build GRCm38 | |
| Downstream | protocadherin 18 | Pcdh18 |
3110038E07Rik, BB095589, PCDH68L |
387.3kb | Anti-sense | Chr. 3 - 49743291 - 49757382 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm20557
Genomic Map