Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200003593.mm.1 |
NM_001025605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003593.mm.1 |
NM_001025605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003593.mm.1 |
TC1200000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003592.mm.1 |
NM_001025605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003592.mm.1 |
NM_001025605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003592.mm.1 |
TC1200000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003591.mm.1 |
NM_001025605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003591.mm.1 |
NM_001025605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003591.mm.1 |
TC1200000521.mm.1 |
MTA TC |