Assay Details
Gene Symbol: Gm527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025605.1 | NP_001020776.1 | 4 - 5 | 1032 | 105 | |
GenBank mRNA | BC099503.1 | - | 4 - 5 | 1032 | 30940052 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28492940 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28492939 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28492941 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P288750 | NM_001025605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P288750 | BC099503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P288750 | NM_001025605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P288750 | BC099503.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236153 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445034 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30770 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P288750 | NM_001025605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P288750 | BC099503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P288750 | NM_001025605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P288750 | BC099503.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492934 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492954 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492909 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492926 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492921 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492939 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492933 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492957 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492932 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492936 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28492905 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07645072 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07645084 | - | - |
Whole Mouse Genome G4122A | A_52_P288750 | NM_001025605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288750 | BC099503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288750 | NM_001025605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288750 | BC099503.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07645072 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10363946 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10363940 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10363937 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28492941 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28492939 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28492940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28492957 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492929 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493000 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492889 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492928 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492949 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492926 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09933144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492945 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09933145 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10197871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10250740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492921 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09963006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10197868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492916 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492909 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492986 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493004 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09982597 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493010 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492927 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10363944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492993 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10170914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492957 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28492961 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10395982 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395979 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395980 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395981 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395983 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1439791_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10395978 | - | - |
Mouse Genome 430 2.0 Array | 1439791_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1439791_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217648_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217648_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200003595.mm.1 | NM_001025605.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200003595.mm.1 | NM_001025605.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200003595.mm.1 | TC1200000521.mm.1 | MTA TC |
Mouse Expression Set 430 | 1439791_at | - | - |
Mouse Expression Set 430 | 1439791_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439791_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439791_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 661066 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225573 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3042287 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3117134 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3117134 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3042287 | - | - |
