Assay Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177805.3 | NP_808473.2 | 11 - 12 | 4056 | 98 | |
| GenBank mRNA | AK172948.1 | - | 11 - 12 | 3984 | 98 | |
| BC062107.1 | - | 1 - 2 | 89 | 98 | ||
| BC157958.1 | - | 11 - 12 | 3858 | 9088131 | 98 | |
| BC158008.1 | - | 12 - 13 | 4008 | 9007316 | 98 | |
| BC172117.1 | - | 11 - 12 | 3858 | 9054403 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062107.1 transcripts.
Set Membership: |
|

Fam179b
Genomic Map