Assay Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177805.3 | NP_808473.2 | 16 - 17 | 4797 | 97 | |
| GenBank mRNA | AK148829.1 | - | 4 - 5 | 447 | 97 | |
| AK172948.1 | - | 16 - 17 | 4725 | 97 | ||
| BC062107.1 | - | 6 - 7 | 829 | 97 | ||
| BC157958.1 | - | 16 - 17 | 4599 | 9088131 | 97 | |
| BC158008.1 | - | 17 - 18 | 4749 | 9007316 | 97 | |
| BC172117.1 | - | 16 - 17 | 4599 | 9054403 | 97 | |
More Information
Set Membership: |
|

Fam179b
Genomic Map