Assay Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177805.3 | NP_808473.2 | 18 - 19 | 5205 | 74 | |
| GenBank mRNA | BC062107.1 | - | 8 - 9 | 1237 | 74 | |
More Information
Set Membership: |
|

Fam179b
Genomic Map