Assay Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177805.3 | NP_808473.2 | 5 - 6 | 3144 | 143 | |
| GenBank mRNA | AK141238.1 | - | 5 - 6 | 3084 | 143 | |
| AK172948.1 | - | 5 - 6 | 3072 | 143 | ||
| BC157958.1 | - | 5 - 6 | 2946 | 9088131 | 143 | |
| BC158008.1 | - | 5 - 6 | 2946 | 9007316 | 143 | |
| BC172117.1 | - | 5 - 6 | 2946 | 9054403 | 143 | |
More Information
Set Membership: |
|

Fam179b
Genomic Map