| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200003610.mm.1 |
NM_177805.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003610.mm.1 |
NM_177805.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003610.mm.1 |
TC1200000523.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003606.mm.1 |
NM_177805.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003606.mm.1 |
NM_177805.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003606.mm.1 |
TC1200000523.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003614.mm.1 |
NM_177805.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003614.mm.1 |
NM_177805.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003614.mm.1 |
TC1200000523.mm.1 |
MTA TC |