Assay Details
| Gene Symbol: Intu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175515.5 | NP_780724.3 | 1672 | 93 | ||
| GenBank mRNA | AK122485.1 | - | 9 - 10 | 1212 | 93 | |
| BC125523.1 | - | 1619 | 40130038 | 93 | ||
| BC132387.1 | - | 1619 | 40130664 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Intu
Genomic Map