Assay Details
| Gene Symbol: Canx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110499.1 | NP_001103969.1 | 13 - 14 | 1827 | 117 | |
| NM_001110500.1 | NP_001103970.1 | 13 - 14 | 1795 | 117 | ||
| NM_007597.3 | NP_031623.1 | 13 - 14 | 1831 | 117 | ||
| GenBank mRNA | AK028279.1 | - | 13 - 14 | 1759 | 117 | |
| AK084175.1 | - | 13 - 14 | 1771 | 117 | ||
| AK159298.1 | - | 13 - 14 | 1815 | 117 | ||
| AK165508.1 | - | 13 - 14 | 1730 | 117 | ||
| AK166649.1 | - | 13 - 14 | 1808 | 117 | ||
| BC012408.1 | - | 13 - 14 | 1749 | 3497769 | 117 | |
| BC040244.1 | - | 13 - 14 | 1758 | 2582119 | 117 | |
| L18888.1 | - | 12 - 13 | 1643 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Canx
Genomic Map