Assay Details
| Gene Symbol: Canx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110499.1 | NP_001103969.1 | 7 - 8 | 913 | 152 | |
| NM_001110500.1 | NP_001103970.1 | 7 - 8 | 881 | 152 | ||
| NM_007597.3 | NP_031623.1 | 7 - 8 | 917 | 152 | ||
| GenBank mRNA | AK028279.1 | - | 7 - 8 | 845 | 152 | |
| AK084175.1 | - | 7 - 8 | 857 | 152 | ||
| AK159298.1 | - | 7 - 8 | 901 | 152 | ||
| AK165508.1 | - | 7 - 8 | 816 | 152 | ||
| AK166649.1 | - | 7 - 8 | 894 | 152 | ||
| BC012408.1 | - | 7 - 8 | 835 | 3497769 | 152 | |
| BC040244.1 | - | 7 - 8 | 844 | 2582119 | 152 | |
| L18888.1 | - | 6 - 7 | 729 | 152 | ||
| L23865.1 | - | 6 - 7 | 669 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Canx
Genomic Map