Assay Details
| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145807.1 | NP_001139279.1 | 5 - 6 | 830 | 78 | |
| NM_153539.3 | NP_705767.3 | 5 - 6 | 1188 | 78 | ||
| GenBank mRNA | AK046878.1 | - | 5 - 6 | 1189 | 78 | |
| AK083233.1 | - | 6 - 7 | 879 | 78 | ||
| BC030498.1 | - | 4 - 5 | 806 | 5400263 | 78 | |
| BC099956.1 | - | 5 - 6 | 1165 | 30101722 | 78 | |
More Information
Set Membership: |
|

Brinp3
Genomic Map