Assay Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026435.5 | NP_080711.1 | 4 - 5 | 220 | 87 | |
| GenBank mRNA | AK007788.1 | - | 4 - 5 | 184 | 87 | |
| AK008280.1 | - | 4 - 5 | 183 | 87 | ||
| AK011737.1 | - | 4 - 5 | 201 | 86 | ||
| AK089948.1 | - | 4 - 5 | 186 | 87 | ||
| AK148146.1 | - | 4 - 5 | 190 | 87 | ||
| AK166332.1 | - | 4 - 5 | 214 | 87 | ||
| AK167301.1 | - | 4 - 5 | 189 | 87 | ||
| AK167321.1 | - | 4 - 5 | 184 | 87 | ||
| BC051554.1 | - | 3 - 4 | 184 | 87 | ||
| BC061065.1 | - | 4 - 5 | 219 | 6594260 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ufm1
Genomic Map