Assay Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173382.1 | NP_775558.2 | 6 - 7 | 940 | 87 | |
| GenBank mRNA | AK034174.1 | - | 6 - 7 | 1091 | 87 | |
| AK038404.1 | - | 6 - 7 | 940 | 87 | ||
| AK087909.1 | - | 6 - 7 | 961 | 87 | ||
| AK161440.1 | - | 5 - 6 | 759 | 87 | ||
| BC086674.1 | - | 6 - 7 | 497 | 30653602 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038404.1 transcripts.
Set Membership: |
|

Proser1
Genomic Map