Assay Details
| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011273.2 | NP_035403.1 | 11 - 12 | 1711 | 82 | |
| GenBank mRNA | AF131096.1 | - | 11 - 12 | 1501 | 82 | |
| AF198104.1 | - | 11 - 12 | 1501 | 82 | ||
| AK087140.1 | - | 2 - 3 | 887 | 82 | ||
| AK141660.1 | - | 11 - 12 | 1683 | 82 | ||
| AK147403.1 | - | 11 - 12 | 1579 | 82 | ||
| BC153872.1 | - | 11 - 12 | 1587 | 40110725 | 82 | |
| HQ323678.1 | - | 2 - 3 | 352 | 82 | ||
More Information
Set Membership: |
|

Xpr1
Genomic Map