Assay Details
| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011273.2 | NP_035403.1 | 12 - 13 | 1884 | 148 | |
| GenBank mRNA | AF131096.1 | - | 12 - 13 | 1674 | 148 | |
| AF131102.1 | - | 12 - 13 | 1674 | 148 | ||
| AF198104.1 | - | 12 - 13 | 1674 | 148 | ||
| AK087140.1 | - | 3 - 4 | 1060 | 148 | ||
| AK141660.1 | - | 12 - 13 | 1856 | 148 | ||
| AK147403.1 | - | 12 - 13 | 1752 | 148 | ||
| BC153872.1 | - | 12 - 13 | 1760 | 40110725 | 148 | |
| HQ022845.1 | - | 12 - 13 | 1674 | 148 | ||
| HQ022853.1 | - | 12 - 13 | 1669 | 148 | ||
| HQ323678.1 | - | 3 - 4 | 525 | 148 | ||
| HQ323679.1 | - | 12 - 13 | 1674 | 148 | ||
| HQ323680.1 | - | 12 - 13 | 1674 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xpr1
Genomic Map