Assay Details
| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011273.2 | NP_035403.1 | 5 - 6 | 805 | 95 | |
| GenBank mRNA | AF114753.1 | - | 5 - 6 | 690 | 95 | |
| AF131096.1 | - | 5 - 6 | 595 | 95 | ||
| AF131098.1 | - | 5 - 6 | 595 | 95 | ||
| AF131102.1 | - | 5 - 6 | 595 | 95 | ||
| AF198104.1 | - | 5 - 6 | 595 | 95 | ||
| AK054130.1 | - | 1 - 2 | 95 | 95 | ||
| AK141660.1 | - | 5 - 6 | 777 | 95 | ||
| AK147403.1 | - | 5 - 6 | 673 | 95 | ||
| BC075721.1 | - | 5 - 6 | 2537 | 95 | ||
| BC153872.1 | - | 5 - 6 | 681 | 40110725 | 95 | |
| HQ022845.1 | - | 5 - 6 | 595 | 95 | ||
| HQ022853.1 | - | 5 - 6 | 590 | 95 | ||
| HQ323679.1 | - | 5 - 6 | 595 | 95 | ||
| HQ323680.1 | - | 5 - 6 | 595 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xpr1
Genomic Map