Assay Details
| Gene Symbol: Rasal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177644.5 | NP_808312.3 | 14 - 15 | 3924 | 92 | |
| GenBank mRNA | BC120902.2 | - | 14 - 15 | 3267 | 40086771 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

Rasal2
Genomic Map