Assay Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111051.1 | NP_001104521.1 | 10 - 11 | 1242 | 76 | |
| NM_001111052.1 | NP_001104522.1 | 10 - 11 | 1242 | 76 | ||
| NM_001195538.1 | NP_001182467.1 | 14 - 15 | 2103 | 76 | ||
| NM_001195539.1 | NP_001182468.1 | 10 - 11 | 1290 | 76 | ||
| NM_019978.3 | NP_064362.1 | 14 - 15 | 2151 | 76 | ||
| GenBank mRNA | AB093234.1 | - | 14 - 15 | 2057 | 76 | |
| AF155819.1 | - | 14 - 15 | 2049 | 76 | ||
| AF155821.1 | - | 10 - 11 | 1101 | 76 | ||
| AK032424.1 | - | 10 - 11 | 1132 | 76 | ||
| AK047716.1 | - | 10 - 11 | 1029 | 76 | ||
| AK081200.1 | - | 10 - 11 | 1085 | 76 | ||
| AY968047.1 | - | 10 - 11 | 990 | 76 | ||
| AY968048.1 | - | 10 - 11 | 993 | 76 | ||
| BC133685.1 | - | 14 - 15 | 1905 | 40110683 | 76 | |
| BQ930085.1 | - | 2 - 3 | 178 | 76 | ||
More Information
Set Membership: |
|

Dclk1
Genomic Map