Assay Details
| Gene Symbol: Taf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020614.2 | NP_065639.2 | 12 - 13 | 1317 | 120 | |
| NR_028341.1 | 11 - 12 | 1219 | 120 | |||
| GenBank mRNA | AK131963.1 | - | 12 - 13 | 1291 | 120 | |
| AK165580.1 | - | 12 - 13 | 1297 | 120 | ||
| AK170313.1 | - | 12 - 13 | 1283 | 120 | ||
| BC057167.1 | - | 12 - 13 | 1304 | 4954753 | 120 | |
| BC069907.1 | - | 11 - 12 | 1174 | 4951630 | 120 | |
| BC086643.1 | - | 12 - 13 | 1278 | 120 | ||
| BC094035.1 | - | 11 - 12 | 1169 | 5029813 | 120 | |
| BC125027.1 | - | 12 - 13 | 1266 | 40061781 | 120 | |
| BC125028.1 | - | 12 - 13 | 1266 | 40061782 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf1b
Genomic Map