Assay Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199021.3 | NP_950186.3 | 9 - 10 | 984 | 106 | |
| GenBank mRNA | AK032579.1 | - | 7 - 8 | 630 | 106 | |
| AK046842.1 | - | 9 - 10 | 985 | 106 | ||
| BC067026.1 | - | 9 - 10 | 977 | 6853947 | 106 | |
More Information
Set Membership: |
|

Dpp10
Genomic Map