Assay Details
| Gene Symbol: Mme | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289462.1 | NP_001276391.1 | 20 - 21 | 2435 | 87 | |
| NM_001289463.1 | NP_001276392.1 | 20 - 21 | 2219 | 87 | ||
| NM_008604.4 | NP_032630.2 | 20 - 21 | 2069 | 87 | ||
| GenBank mRNA | AK031446.1 | - | 20 - 21 | 2070 | 87 | |
| AK033824.1 | - | 20 - 21 | 2130 | 87 | ||
| AK076908.1 | - | 20 - 21 | 2283 | 87 | ||
| BC034092.1 | - | 20 - 21 | 2091 | 4483558 | 87 | |
| BC066840.1 | - | 19 - 20 | 1978 | 87 | ||
| M81591.1 | - | 19 - 20 | 1981 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mme
Genomic Map