Assay Details
| Gene Symbol: Zcchc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122675.1 | NP_001116147.1 | 6 - 7 | 1762 | 125 | |
| NM_001122676.1 | NP_001116148.1 | 6 - 7 | 1762 | 125 | ||
| GenBank mRNA | AK046562.1 | - | 6 - 7 | 1763 | 125 | |
| AK173248.1 | - | 6 - 7 | 1479 | 125 | ||
| BC035528.1 | - | 3 - 4 | 252 | 125 | ||
More Information
Set Membership: |
|

Zcchc2
Genomic Map