Assay Details
| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081386.1 | NP_001074855.1 | 8 - 9 | 1561 | 68 | |
| GenBank mRNA | AK148396.1 | - | 8 - 9 | 1549 | 68 | |
| BC157927.1 | - | 8 - 9 | 1552 | 9088100 | 68 | |
More Information
Set Membership: |
|

Cdh19
Genomic Map