Assay Details
| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081307.1 | NP_001074776.1 | 13 - 14 | 2001 | 82 | |
| GenBank mRNA | AK141375.1 | - | 15 - 16 | 2216 | 82 | |
| BC145746.1 | - | 13 - 14 | 1912 | 40131049 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r12b
Genomic Map