Assay Details
| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081307.1 | NP_001074776.1 | 19 - 20 | 2648 | 120 | |
| GenBank mRNA | AK039445.1 | - | 7 - 8 | 709 | 120 | |
| BC145746.1 | - | 19 - 20 | 2559 | 40131049 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r12b
Genomic Map