Assay Details
| Gene Symbol: Hjurp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198652.2 | NP_941054.2 | 4 - 5 | 365 | 114 | |
| GenBank mRNA | AB162219.1 | - | 4 - 5 | 255 | 114 | |
| BC057309.1 | - | 4 - 5 | 306 | 6826508 | 114 | |
More Information
Set Membership: |
|

Hjurp
Genomic Map