Assay Details
| Gene Symbol: Hjurp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198652.2 | NP_941054.2 | 5 - 6 | 462 | 111 | |
| GenBank mRNA | AB162219.1 | - | 5 - 6 | 352 | 111 | |
| BC057309.1 | - | 5 - 6 | 403 | 6826508 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045096 | BC057309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045096 | NM_198652.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045096 | AB162219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045096 | AK211133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968200 | BC057309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968200 | NM_198652.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968200 | AB162219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968200 | BC062125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968200 | AK032612.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P01247 | BC062125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P01247 | AK032612.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P01247 | AK135880.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871981 | BC099526.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871981 | XM_001473995.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 321033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106659 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142183 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169507 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103858 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045096 | BC057309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045096 | NM_198652.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045096 | AB162219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045096 | AK211133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968200 | BC057309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968200 | NM_198652.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968200 | AB162219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968200 | BC062125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968200 | AK032612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P01247 | BC062125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P01247 | AK032612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P01247 | AK135880.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20422693 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04179655 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142183 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169507 | - | - |
| Whole Mouse Genome G4122A | A_52_P221554 | BC004768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P221554 | BC057309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P221554 | NM_198652.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P221554 | AB162219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P449624 | BC004768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P957031 | NM_001177364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P957031 | NM_001177364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50474 | BC057309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50474 | NM_198652.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50474 | BC062125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50474 | AK135880.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140025 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139270 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169507 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20422612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01064197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00880305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00472410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00472408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00781376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00472422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00472409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00472415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00962667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00199432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00880307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00947070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00182677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00947073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20422504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10356465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356468 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356463 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442202_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451456_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455654_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10356461 | - | - |
| Mouse Genome 430 2.0 Array | 1451456_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433685_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456631_at | - | - |
| Mouse Genome 430 2.0 Array | 1447368_at | - | - |
| Mouse Genome 430 2.0 Array | 1454686_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455654_at | - | - |
| Mouse Genome 430 2.0 Array | 1442202_at | - | - |
| Mouse Genome 430 2.0 Array | 1442202_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456631_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454686_at | - | - |
| Mouse Genome 430 2.0 Array | 1447368_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451456_at | - | - |
| Mouse Genome 430 2.0 Array | 1433685_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455654_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381280_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381280_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100022400.mm.1 | NM_198652.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022400.mm.1 | NM_198652.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022400.mm.1 | TC0100002772.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100022386.mm.1 | NM_198652.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022386.mm.1 | NM_198652.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022386.mm.1 | TC0100002772.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451456_at | - | - |
| Mouse Expression Set 430 | 1433685_a_at | - | - |
| Mouse Expression Set 430 | 1454686_PM_at | - | - |
| Mouse Expression Set 430 | 1433685_PM_a_at | - | - |
| Mouse Expression Set 430 | 1454686_at | - | - |
| Mouse Expression Set 430 | 1451456_PM_at | - | - |
| Mouse Expression Set 430 | 1447368_at | - | - |
| Mouse Expression Set 430 | 1455654_PM_at | - | - |
| Mouse Expression Set 430 | 1442202_at | - | - |
| Mouse Expression Set 430 | 1455654_at | - | - |
| Mouse Expression Set 430 | 1447368_PM_at | - | - |
| Mouse Expression Set 430 | 1442202_PM_at | - | - |
| Murine Genome U74v2 Set | 160682_PM_at | - | - |
| Murine Genome U74v2 Set | 160682_at | - | - |
| Murine Genome U74v2 Set | 128809_at | - | - |
| Murine Genome U74v2 Set | 128809_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451456_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454686_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454686_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433685_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451456_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433685_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451456_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451456_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455654_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442202_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442202_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455654_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_3011719 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3011719 | - | - |

Hjurp
Genomic Map