Assay Details
| Gene Symbol: Nav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173437.2 | NP_775613.2 | 21 - 22 | 4331 | 110 | |
| GenBank mRNA | AF307453.1 | - | 21 - 22 | 4691 | 110 | |
| AK036761.1 | - | 3 - 4 | 335 | 110 | ||
| AK122458.1 | - | 21 - 22 | 4389 | 110 | ||
| AK153742.1 | - | 4 - 5 | 652 | 110 | ||
| BC080292.1 | - | 3 - 4 | 403 | 110 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK147844.1 | - | Chr. - - [-] on Build | 110 | |||
More Information
Set Membership: |
|

Nav1
Genomic Map