Assay Details
| Gene Symbol: Dhx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007842.2 | NP_031868.2 | 25 - 26 | 3304 | 84 | |
| GenBank mRNA | AK169239.1 | - | 1 - 2 | 181 | 84 | |
| CB181391.1 | - | 3 - 4 | 435 | 84 | ||
| U91922.1 | - | 25 - 26 | 3261 | 84 | ||
More Information
Set Membership: |
|

Dhx9
Genomic Map