Assay Details
| Gene Symbol: Rgsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243223.1 | NP_001230152.1 | 12 - 13 | 2270 | 78 | |
| GenBank mRNA | AK029630.1 | - | 3 - 4 | 377 | 78 | |
| AK161463.1 | - | 5 - 6 | 687 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgsl1
Genomic Map