Assay Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010049.3 | NP_034179.1 | 3 - 4 | 724 | 134 | |
| GenBank mRNA | AK031241.1 | - | 3 - 4 | 358 | 134 | |
| AK136212.1 | - | 3 - 4 | 782 | 134 | ||
| BC005796.1 | - | 3 - 4 | 296 | 3707582 | 134 | |
| L26316.1 | - | 3 - 4 | 303 | 134 | ||
| M10811.1 | - | 1 - 2 | 101 | 134 | ||
| V00731.1 | - | 381 | 134 | |||
| V00734.1 | - | 3 - 4 | 360 | 134 | ||
| X56066.1 | - | 3 - 4 | 270 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhfr
Genomic Map