Assay Details
| Gene Symbol: Msh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010829.2 | NP_034959.2 | 5 - 6 | 883 | 63 | |
| GenBank mRNA | AK011426.1 | - | 6 - 7 | 1486 | 63 | |
| AK040616.1 | - | 4 - 5 | 668 | 63 | ||
| AK078467.1 | - | 5 - 6 | 834 | 63 | ||
| AK083388.1 | - | 5 - 6 | 789 | 63 | ||
| AK133907.1 | - | 5 - 6 | 875 | 63 | ||
| AK161089.1 | - | 5 - 6 | 879 | 63 | ||
| BC150759.1 | - | 5 - 6 | 802 | 9087670 | 63 | |
| M24919.1 | - | 5 - 6 | 1166 | 63 | ||
| M80360.1 | - | 5 - 6 | 880 | 63 | ||
More Information
Set Membership: |
|

Msh3
Genomic Map