Assay Details
| Gene Symbol: Ckmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198415.2 | NP_940807.1 | 3 - 4 | 442 | 79 | |
| GenBank mRNA | AK009042.1 | - | 1 - 2 | 146 | 79 | |
| BC061221.1 | - | 3 - 4 | 442 | 30296538 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ckmt2
Genomic Map