Assay Details
| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167949.2 | NP_001161421.1 | 13 - 14 | 2764 | 78 | |
| NM_213616.4 | NP_998781.1 | 13 - 14 | 2764 | 78 | ||
| GenBank mRNA | AY560895.1 | - | 13 - 14 | 2298 | 78 | |
| BC109172.1 | - | 13 - 14 | 2298 | 40048499 | 78 | |
| BC109173.1 | - | 13 - 14 | 2298 | 40048500 | 78 | |
| EU493094.2 | - | 13 - 14 | 2468 | 78 | ||
| EU493095.2 | - | 13 - 14 | 2468 | 78 | ||
| JF683384.1 | - | 13 - 14 | 2468 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2b4
Genomic Map