Assay Details
| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167949.2 | NP_001161421.1 | 9 - 10 | 2022 | 66 | |
| NM_213616.4 | NP_998781.1 | 9 - 10 | 2022 | 66 | ||
| GenBank mRNA | AY560895.1 | - | 9 - 10 | 1556 | 66 | |
| BC109172.1 | - | 9 - 10 | 1556 | 40048499 | 66 | |
| BC109173.1 | - | 9 - 10 | 1556 | 40048500 | 66 | |
| EU493094.2 | - | 9 - 10 | 1726 | 66 | ||
| EU493095.2 | - | 9 - 10 | 1726 | 66 | ||
| JF683384.1 | - | 9 - 10 | 1726 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2b4
Genomic Map