Assay Details
| Gene Symbol: Per2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011066.3 | NP_035196.2 | 5 - 6 | 736 | 70 | |
| GenBank mRNA | AF035830.1 | - | 5 - 6 | 683 | 70 | |
| AF036893.1 | - | 5 - 6 | 706 | 70 | ||
| AK044658.1 | - | 5 - 6 | 681 | 70 | ||
| AK122253.1 | - | 5 - 6 | 701 | 70 | ||
| AK159847.1 | - | 5 - 6 | 738 | 70 | ||
| AK165556.1 | - | 5 - 6 | 739 | 70 | ||
More Information
Set Membership: |
|

Per2
Genomic Map