Assay Details
| Gene Symbol: Traf3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028718.2 | NP_082994.1 | 9 - 10 | 1262 | 100 | |
| GenBank mRNA | AK014457.1 | - | 6 - 7 | 743 | 100 | |
| AY613437.1 | - | 9 - 10 | 1265 | 100 | ||
| BC051115.1 | - | 2 - 3 | 136 | 100 | ||
| BC117868.2 | - | 9 - 10 | 1236 | 40095476 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Traf3ip1
Genomic Map