| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300001866.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001866.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001866.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001867.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001867.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001867.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001872.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001872.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001872.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001870.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001870.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001870.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001869.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001869.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001869.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001871.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001871.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001871.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001873.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001873.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001873.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001868.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001868.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001868.mm.1 |
TC0300000272.mm.1 |
MTA TC |