Assay Details
| Gene Symbol: 4932438A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172679.2 | NP_766267.2 | 72 - 73 | 12726 | 59 | |
| GenBank mRNA | AK052702.1 | - | 4 - 5 | 544 | 59 | |
| AK086976.1 | - | 7 - 8 | 2925 | 59 | ||
| AK173100.1 | - | 16 - 17 | 2361 | 59 | ||
| BC079623.1 | - | 13 - 14 | 2096 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

4932438A13Rik
Genomic Map